Learn More
Chemglass Life Sciences Large Diameter Sampling Tube/ Harvest Tube, M18, 2-5 Liter BioReactors
SDP
Catalog No. 501532114
Encompass
Pipe will fit a 2 to 5 liter reactor and has a lower diameter of 1/2" and a length of 212mm into the reactor.The serrated hose connection is also 1/2" (12.7mm) OD.Can also be used as a quick way to harvest material due to its size.Stainless steel components are manufactured from 316L with an electropolished surface finish of 20Ra or better.
This item is not returnable. View return policy
Catalog No. 50-153-2114 Supplier Chemglass Life Sciences Supplier No. CLS138070
Pipe will fit a 2 to 5 liter reactor and has a lower diameter of 1/2" and a length of 212mm into the reactor.The serrated hose connection is also 1/2" (12.7mm) OD.Can also be used as a quick way to harvest material due to its size.Stainless steel components are manufactured from 316L with an electropolished surface finish of 20Ra or better.