Learn More
Cayman Chemical LArgInIn13C6hydrochLRId 10mg
SDP
Catalog No. 502894561
Encompass_Preferred
An internal standard for the quantification of L-arginine by GC- or LC-MS
Catalog No. 50-289-4561 Supplier Cayman Chemical Supplier No. 900408810 MG
An internal standard for the quantification of L-arginine by GC- or LC-MS