Learn More
Abcam Lariatin A, antimycobacterial agent, 1MG
Catalog No. 502705918
Encompass_Preferred
MW 2036.3 Da, Purity >95%. Potent antimycobacterial agent. Contains a lasso structure. Lariatin B analog. Inhibits the growth of Mycobacterium smegmatis (MIC = 3 μM) and Mycobacterium tuberculosis (MIC = 0.2 μM). Shows antimycobacterial effects in vivo. .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5918 Supplier Abcam Supplier No. AB1442881MG
MW 2036.3 Da, Purity >95%. Potent antimycobacterial agent. Contains a lasso structure. Lariatin B analog. Inhibits the growth of Mycobacterium smegmatis (MIC = 3 μM) and Mycobacterium tuberculosis (MIC = 0.2 μM). Shows antimycobacterial effects in vivo. .
The product is subject to the following: Abcam Restricted Use Statement