Learn More
Medchemexpress LLC Larixyl acetate | 4608-49-5 | 99.5% | 348.52 | C22H36O3 | 25 MG
SDP
Catalog No. 5000248898
Encompass_Preferred
Larixyl acetate is a diterpenoid research compound that inhibits TRPC6 channels and is used in cellular and animal studies of ion channel signaling, inflammation, cardiac hypertrophy, and traumatic brain injury. It is supplied as a powder (formula C22H36O3; MW 348.52 g/mol) and is intended for research use only.
  • Potent TRPC6 inhibitor (IC50 0.58 μM).
  • Also active against TRPC3 (IC50 6.83 μM).
  • High purity (99.5%).
  • Supplied as powder with multiple pack sizes, including 25 MG.
  • Recommended storage: powder at -20°C; solutions at -80°C for long-term.
  • Suitable for in vitro and in vivo pharmacology studies.
Catalog No. 50-002-48898 Supplier Medchemexpress LLC Supplier No. HY10179525MG
Larixyl acetate is a diterpenoid research compound that inhibits TRPC6 channels and is used in cellular and animal studies of ion channel signaling, inflammation, cardiac hypertrophy, and traumatic brain injury. It is supplied as a powder (formula C22H36O3; MW 348.52 g/mol) and is intended for research use only.
  • Potent TRPC6 inhibitor (IC50 0.58 μM).
  • Also active against TRPC3 (IC50 6.83 μM).
  • High purity (99.5%).
  • Supplied as powder with multiple pack sizes, including 25 MG.
  • Recommended storage: powder at -20°C; solutions at -80°C for long-term.
  • Suitable for in vitro and in vivo pharmacology studies.