Learn More
Medchemexpress LLC Larixyl acetate | 4608-49-5 | 99.5% | 348.52 g·mol⁻¹ | C22H36O3 | 1 ML
SDP
Catalog No. 5000248172
Encompass_Preferred
Larixyl acetate is a diterpenoid research reagent that acts as a potent, selective TRPC6 inhibitor. Supplied as a 10 mM solution in DMSO, it is intended for laboratory pharmacology and biochemical studies targeting TRPC channels and related pathways.
  • Potent, selective TRPC6 inhibition demonstrated in cellular assays.
  • Provided as a ready-to-use 10 mM solution for convenient dosing.
  • High purity suitable for analytical and research applications.
  • Well-characterized molecular weight (348.52 g·mol⁻¹) for dosing calculations.
  • Storage recommendations available for both powder and solution to preserve stability.
Catalog No. 50-002-48172 Supplier Medchemexpress LLC Supplier No. HY10179510MM 1ML
Larixyl acetate is a diterpenoid research reagent that acts as a potent, selective TRPC6 inhibitor. Supplied as a 10 mM solution in DMSO, it is intended for laboratory pharmacology and biochemical studies targeting TRPC channels and related pathways.
  • Potent, selective TRPC6 inhibition demonstrated in cellular assays.
  • Provided as a ready-to-use 10 mM solution for convenient dosing.
  • High purity suitable for analytical and research applications.
  • Well-characterized molecular weight (348.52 g·mol⁻¹) for dosing calculations.
  • Storage recommendations available for both powder and solution to preserve stability.