Learn More
Medchemexpress LLC Larixyl acetate | 4608-49-5 | 99.5% | 348.52 g/mol | C22H36O3 | 100 MG
SDP
Catalog No. 5000248604
Encompass_Preferred
Larixyl acetate is a diterpenoid research chemical used as an analytical standard and a tool compound in pharmacology studies. It acts as a potent, selective TRPC6 inhibitor with reported hTRPC6 IC50 = 0.58 μM and activity against hTRPC3. Supplied as a solid or as a 10 mM solution in DMSO, it is intended for research use only and requires low-temperature storage to preserve stability.
  • Potent TRPC6 inhibitory activity (hTRPC6 IC50 0.58 μM).
  • Active against related TRPC channels (hTRPC3 IC50 6.83 μM).
  • High purity (~99.5%) suitable for analytical applications.
  • Available as solid and as a 10 mM solution in DMSO, multiple pack sizes including 100 MG.
  • Stable as powder at -20°C for up to 3 years; in solvent store at -80°C (6 months) or -20°C (1 month).
  • Intended for laboratory research use only; not for clinical or human use.
Catalog No. 50-002-48604 Supplier Medchemexpress LLC Supplier No. HY101795100MG
Larixyl acetate is a diterpenoid research chemical used as an analytical standard and a tool compound in pharmacology studies. It acts as a potent, selective TRPC6 inhibitor with reported hTRPC6 IC50 = 0.58 μM and activity against hTRPC3. Supplied as a solid or as a 10 mM solution in DMSO, it is intended for research use only and requires low-temperature storage to preserve stability.
  • Potent TRPC6 inhibitory activity (hTRPC6 IC50 0.58 μM).
  • Active against related TRPC channels (hTRPC3 IC50 6.83 μM).
  • High purity (~99.5%) suitable for analytical applications.
  • Available as solid and as a 10 mM solution in DMSO, multiple pack sizes including 100 MG.
  • Stable as powder at -20°C for up to 3 years; in solvent store at -80°C (6 months) or -20°C (1 month).
  • Intended for laboratory research use only; not for clinical or human use.