Learn More
Cayman Chemical Larixyl Acetate, Purity: ≥98%, CAS: 4608-49-5, 50 mg
SDP
Catalog No. 502913482
Encompass_Preferred
A diterpenoid; inhibits the activity of TRPC6 (IC50 = 0.58 µM receptor) and is selective for TRPC6 over TRPC3 (IC50 = 6.83 µM), as well as over nine additional TRP channels at 10 µM; reduces mechanical and cold allodynia in a rat model of neuropathic pain induced by SNI
Catalog No. 50-291-3482 Supplier Cayman Chemical Supplier No. 3530950 MG
A diterpenoid; inhibits the activity of TRPC6 (IC50 = 0.58 µM receptor) and is selective for TRPC6 over TRPC3 (IC50 = 6.83 µM), as well as over nine additional TRP channels at 10 µM; reduces mechanical and cold allodynia in a rat model of neuropathic pain induced by SNI