Learn More
Medchemexpress LLC Laropiprant | 571170-77-9 | 99.3% | 435.90 | 100 MG
SDP
Catalog No. 5000364801
Encompass_Preferred
Laropiprant is a potent and selective DP receptor antagonist, exhibiting Ki values of 0.57 nM for the DP receptor and 2.95 nM for the TP Receptor. In vitro studies show Laropiprant acts as an inverse agonist of DP1 cAMP signaling, reducing basal DP1 cAMP levels.
  • Potent and selective DP receptor antagonist
  • Exhibits Ki values of 0.57 nM for DP receptor and 2.95 nM for TP receptor
  • Acts as an inverse agonist of DP1 cAMP signaling in HEK293 cells
  • Promotes DP1 cell surface expression as a pharmacochaperone
  • Shows good pharmacokinetic profiles in in vivo studies
Catalog No. 50-003-64801 Supplier Medchemexpress LLC Supplier No. 100MGHY50175
Laropiprant is a potent and selective DP receptor antagonist, exhibiting Ki values of 0.57 nM for the DP receptor and 2.95 nM for the TP Receptor. In vitro studies show Laropiprant acts as an inverse agonist of DP1 cAMP signaling, reducing basal DP1 cAMP levels.
  • Potent and selective DP receptor antagonist
  • Exhibits Ki values of 0.57 nM for DP receptor and 2.95 nM for TP receptor
  • Acts as an inverse agonist of DP1 cAMP signaling in HEK293 cells
  • Promotes DP1 cell surface expression as a pharmacochaperone
  • Shows good pharmacokinetic profiles in in vivo studies