Learn More
Medchemexpress LLC HY-125374 1mg Medchemexpress, Larotaxel CAS:156294-36-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1253741MG
Encompass_Preferred
Medchemexpress, HY-125374 1mg Larotaxel CAS:156294-36-9 Larotaxel (XRP9881) is a taxane analogue with preclinical activity against taxane-resistant breast cancer. Larotaxel (XRP9881) exerts its cytotoxic effect by promoting tubulin assembly and stabilizing microtubules, ultimately leading to cell death by apoptosis . It presents the ability to cross the blood brain barrier and has a much lower affinity for P-glycoprotein 1 than Docetaxel [1] [2] [3] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2604