Learn More
Medchemexpress LLC Larotinib mesylate hydrate | 2097129-93-4 | C26H36ClFN4O11S2 | 25 MG
SDP
Catalog No. 5000375344
Encompass_Preferred
Larotinib mesylate hydrate is a potent broad-spectrum and orally active tyrosine kinase inhibitor (TKI) with EGFR as the main target with an IC50 of 0.6 nM.
  • Potent broad-spectrum and orally active tyrosine kinase inhibitor (TKI)
  • Primary target is EGFR with an IC50 of 0.6 nM
  • Purity of 98.66%
  • Appears as a white to off-white solid
  • Demonstrates dose-dependent antitumor results in vivo
  • Achieves a tumor-inhibiting rate exceeding 60% in tumor-bearing mice models
Catalog No. 50-003-75344 Supplier Medchemexpress LLC Supplier No. 25MGHY153110A
Larotinib mesylate hydrate is a potent broad-spectrum and orally active tyrosine kinase inhibitor (TKI) with EGFR as the main target with an IC50 of 0.6 nM.
  • Potent broad-spectrum and orally active tyrosine kinase inhibitor (TKI)
  • Primary target is EGFR with an IC50 of 0.6 nM
  • Purity of 98.66%
  • Appears as a white to off-white solid
  • Demonstrates dose-dependent antitumor results in vivo
  • Achieves a tumor-inhibiting rate exceeding 60% in tumor-bearing mice models