Learn More
Medchemexpress LLC Larotrectinib | 1223403-58-4 | 99.9% | 10 MG
SDP
Catalog No. 5000341992
Encompass_Preferred
Larotrectinib is an ATP-competitive oral, selective inhibitor of the tropomyosin-related kinase (TRK) family receptors. It demonstrates low nanomolar 50% inhibitory concentrations against all three isoforms (TRKA, B, and C) and exhibits 1,000-fold or greater selectivity relative to other kinases. This product is for research use only.
  • Targets TrkA, TrkB, and TrkC
  • Exhibits antiproliferative activity against various cell lines
  • Demonstrates dose-dependent inhibition of cell proliferation
  • Has low brain penetration in animal studies
  • Well tolerated in GLP toxicology studies
  • Reduces tyrosine phosphorylation of TRKA and downstream signal transduction in tumors
  • Shows dose-dependent tumor inhibition in athymic nude mice
  • Reduces leukemic infiltration in bone marrow and spleen in mice
Catalog No. 50-003-41992 Supplier Medchemexpress LLC Supplier No. HY1286610MG
Larotrectinib is an ATP-competitive oral, selective inhibitor of the tropomyosin-related kinase (TRK) family receptors. It demonstrates low nanomolar 50% inhibitory concentrations against all three isoforms (TRKA, B, and C) and exhibits 1,000-fold or greater selectivity relative to other kinases. This product is for research use only.
  • Targets TrkA, TrkB, and TrkC
  • Exhibits antiproliferative activity against various cell lines
  • Demonstrates dose-dependent inhibition of cell proliferation
  • Has low brain penetration in animal studies
  • Well tolerated in GLP toxicology studies
  • Reduces tyrosine phosphorylation of TRKA and downstream signal transduction in tumors
  • Shows dose-dependent tumor inhibition in athymic nude mice
  • Reduces leukemic infiltration in bone marrow and spleen in mice