Learn More
Selleck Chemical LLC Larotrectinib-5mg
SDP
Catalog No. 502389846
Encompass_Preferred
Larotrectinib (Vitrakvi, LOXO-101, ARRY-470) is an ATP-competitive, oral administered and highly selective inhibitor of the tropomyosin-related kinase (TRK) family receptors.
Catalog No. 50-238-9846 Supplier Selleck Chemical LLC Supplier No. S58605MG
Larotrectinib (Vitrakvi, LOXO-101, ARRY-470) is an ATP-competitive, oral administered and highly selective inhibitor of the tropomyosin-related kinase (TRK) family receptors.