Learn More
Selleck Chemical LLC Larotrectinib
SDP
Catalog No. 5000003632
Encompass_Preferred
Larotrectinib (Vitrakvi LOXO-101 ARRY-470) is an ATP-competitive oral administered and highly selective inhibitor of the tropomyosin-related kinase (TRK) family receptors
Catalog No. 50-000-03632 Supplier Selleck Chemical LLC Supplier No. S58601G
Larotrectinib (Vitrakvi LOXO-101 ARRY-470) is an ATP-competitive oral administered and highly selective inhibitor of the tropomyosin-related kinase (TRK) family receptors