Learn More
Aviva Systems Biology Corp LARP1 Antibody - N-terminal region (ARP41077_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41077_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LARP1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human LARP1. Alternate names: KIAA0731, LARP, MGC19556ApplicationWB. Species reactivity: Human, Mouse, Rabbit
Catalog No. 50-284-911