Learn More
Aviva Systems Biology Corp LARP6 Antibody - middle region (ARP40912_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40912_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LARP6. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human LARP6. Alternate names: ACHN, FLJ11196ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-57660