Learn More
Aviva Systems Biology Corp LARP7 Antibody - C-terminal region (ARP40847_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40847_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LARP7. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human LARP7. Alternate names: DKFZP564K112, HDCMA18P, MGC104360, PIP7SApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-53244