Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Larsucosterol (trimethylamine) | 00-00-0 | 97.0% | 509.32 | C27H46O5S · 0.45 C3H9N | 5 MG
SDP

Catalog No. 5000222216
Encompass_Preferred

Larsucosterol (trimethylamine) is a research-grade small molecule reported as an LXR antagonist and epigenetic modulator. Supplied as the trimethylamine salt, it is intended for biochemical and pharmacological studies of cholesterol metabolism and related pathways. Available as a solid and as a DMSO solution for in vitro research use only.

  • High purity (97.0%) suitable for research applications.
  • Molecular weight 509.32 with formula C27H46O5S · 0.45 C3H9N.
  • Available in small pack sizes including 5 mg for screening and characterization.
  • Solid storage: 4°C, sealed and protected from moisture.
  • Solution stability: -80°C for up to 6 months, -20°C for up to 1 month.
  • Used to study LXR-related signaling and cholesterol metabolism pathways.

Catalog No. 50-002-22216 Supplier Medchemexpress LLC Supplier No. HY139576B5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Larsucosterol (trimethylamine) is a research-grade small molecule reported as an LXR antagonist and epigenetic modulator. Supplied as the trimethylamine salt, it is intended for biochemical and pharmacological studies of cholesterol metabolism and related pathways. Available as a solid and as a DMSO solution for in vitro research use only.

  • High purity (97.0%) suitable for research applications.
  • Molecular weight 509.32 with formula C27H46O5S · 0.45 C3H9N.
  • Available in small pack sizes including 5 mg for screening and characterization.
  • Solid storage: 4°C, sealed and protected from moisture.
  • Solution stability: -80°C for up to 6 months, -20°C for up to 1 month.
  • Used to study LXR-related signaling and cholesterol metabolism pathways.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.