Learn More
Medchemexpress LLC Larsucosterol trimethylamine | 00-00-0 | 97.0% | C27H46O5S·0.45C3H9N | 10MG
SDP
Catalog No. 5000261672
Encompass_Preferred
Larsucosterol trimethylamine is a cholesterol metabolite used as a research reagent. It acts as a potent liver X receptor (LXR) antagonist and modulates lipid metabolism by decreasing lipogenesis and inhibiting cholesterol biosynthesis through reduced SREBP-1 activation. The compound is supplied in high-purity forms for laboratory assays, available as small solid quantities or ready-to-use DMSO solutions.
  • Potent liver X receptor (LXR) antagonist
  • Modulates lipid metabolism by decreasing lipogenesis
  • Inhibits cholesterol biosynthesis via reduced SREBP-1 activation
  • High purity (97.0%) suitable for research use
  • Available in small solid quantities and DMSO solutions for assays
Catalog No. 50-002-61672 Supplier Medchemexpress LLC Supplier No. HY139576B10MG
Larsucosterol trimethylamine is a cholesterol metabolite used as a research reagent. It acts as a potent liver X receptor (LXR) antagonist and modulates lipid metabolism by decreasing lipogenesis and inhibiting cholesterol biosynthesis through reduced SREBP-1 activation. The compound is supplied in high-purity forms for laboratory assays, available as small solid quantities or ready-to-use DMSO solutions.
  • Potent liver X receptor (LXR) antagonist
  • Modulates lipid metabolism by decreasing lipogenesis
  • Inhibits cholesterol biosynthesis via reduced SREBP-1 activation
  • High purity (97.0%) suitable for research use
  • Available in small solid quantities and DMSO solutions for assays