Learn More
Medchemexpress LLC Larsucosterol (trimethylamine) | 2655654-16-1 | 97.0% | 509.32 | 1 MG
SDP
Catalog No. 5000350398
Encompass_Preferred
Larsucosterol (DUR-928) trimethylamine is a cholesterol metabolite and a potent liver X receptor (LXR) antagonist. This compound acts as an endogenous regulator to decrease lipogenesis, inhibiting cholesterol biosynthesis by decreasing mRNA levels and blocking the activation of SREBP-1.
  • Potent liver X receptor (LXR) antagonist.
  • Decreases lipogenesis.
  • Inhibits cholesterol biosynthesis.
  • Blocks SREBP-1 activation.
  • Reduces serum lipid levels in NAFLD models.
  • Suppresses hepatic inflammation.
Catalog No. 50-003-50398 Supplier Medchemexpress LLC Supplier No. HY139576B1MG
Larsucosterol (DUR-928) trimethylamine is a cholesterol metabolite and a potent liver X receptor (LXR) antagonist. This compound acts as an endogenous regulator to decrease lipogenesis, inhibiting cholesterol biosynthesis by decreasing mRNA levels and blocking the activation of SREBP-1.
  • Potent liver X receptor (LXR) antagonist.
  • Decreases lipogenesis.
  • Inhibits cholesterol biosynthesis.
  • Blocks SREBP-1 activation.
  • Reduces serum lipid levels in NAFLD models.
  • Suppresses hepatic inflammation.