Learn More
Medchemexpress LLC Larsucosterol (trimethylamine) | 97.0% | 509.32 | 1 ML
SDP
Catalog No. 5000350397
Encompass_Preferred
Larsucosterol (trimethylamine), also known as DUR-928 (trimethylamine), is a cholesterol metabolite that functions as a potent liver X receptor (LXR) antagonist. It serves as an endogenous regulator to decrease lipogenesis and inhibits cholesterol biosynthesis by reducing mRNA levels and inhibiting the activation of SREBP-1.
  • Potent liver X receptor (LXR) antagonist
  • Decreases lipogenesis
  • Inhibits cholesterol biosynthesis by reducing mRNA levels and activating SREBP-1
  • Increases cell proliferation and decreases apoptosis in macrophages
  • Protects the liver from injury by suppressing hepatic inflammation
Catalog No. 50-003-50397 Supplier Medchemexpress LLC Supplier No. HY139576B10MM/1ML
Larsucosterol (trimethylamine), also known as DUR-928 (trimethylamine), is a cholesterol metabolite that functions as a potent liver X receptor (LXR) antagonist. It serves as an endogenous regulator to decrease lipogenesis and inhibits cholesterol biosynthesis by reducing mRNA levels and inhibiting the activation of SREBP-1.
  • Potent liver X receptor (LXR) antagonist
  • Decreases lipogenesis
  • Inhibits cholesterol biosynthesis by reducing mRNA levels and activating SREBP-1
  • Increases cell proliferation and decreases apoptosis in macrophages
  • Protects the liver from injury by suppressing hepatic inflammation