Learn More
Medchemexpress LLC Larsucosterol (trimethylamine) | 97.0% | 509.32 | 25 MG
SDP
Catalog No. 5000349951
Encompass_Preferred
Larsucosterol (trimethylamine) is a cholesterol metabolite that functions as a potent liver X receptor (LXR) antagonist. This endogenous regulator decreases lipogenesis and inhibits cholesterol biosynthesis by reducing mRNA levels and inhibiting the activation of SREBP-1.
  • Potent liver X receptor (LXR) antagonist
  • Decreases lipogenesis
  • Inhibits cholesterol biosynthesis
  • Reduces mRNA levels of cholesterol biosynthesis enzymes
  • Inhibits activation of SREBP-1
  • Increases cell proliferation in macrophages
  • Decreases apoptosis in macrophages
  • Reduces serum lipid levels in NAFLD models
  • Protects liver from injury by suppressing hepatic inflammation
Catalog No. 50-003-49951 Supplier Medchemexpress LLC Supplier No. 25MGHY139576B
Larsucosterol (trimethylamine) is a cholesterol metabolite that functions as a potent liver X receptor (LXR) antagonist. This endogenous regulator decreases lipogenesis and inhibits cholesterol biosynthesis by reducing mRNA levels and inhibiting the activation of SREBP-1.
  • Potent liver X receptor (LXR) antagonist
  • Decreases lipogenesis
  • Inhibits cholesterol biosynthesis
  • Reduces mRNA levels of cholesterol biosynthesis enzymes
  • Inhibits activation of SREBP-1
  • Increases cell proliferation in macrophages
  • Decreases apoptosis in macrophages
  • Reduces serum lipid levels in NAFLD models
  • Protects liver from injury by suppressing hepatic inflammation