Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Lartesertib | 2020089-41-0 | 448.45 | C23H21FN6O3 | 10 MG
SDP

Catalog No. 5000286738
Encompass_Preferred

Lartesertib is a potent, small-molecule inhibitor of the ATM (ataxia telangiectasia mutated) serine/threonine protein kinase used in research on DNA damage response and tumor radiosensitization. It is supplied as a research reagent with an associated product datasheet; lot-specific purity is provided on the certificate of analysis.

  • Potent and selective ATM kinase inhibitor suitable for DNA damage response studies.
  • Demonstrated activity in cellular and preclinical models, including radiosensitization.
  • Supplied with a product datasheet; certificate of analysis provides lot-specific purity.
  • Molecular weight 448.45 and molecular formula C23H21FN6O3.
  • CAS number 2020089-41-0; alternate registry entries have been reported in public databases.

Catalog No. 50-002-86738 Supplier Medchemexpress LLC Supplier No. HY15061710MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Lartesertib is a potent, small-molecule inhibitor of the ATM (ataxia telangiectasia mutated) serine/threonine protein kinase used in research on DNA damage response and tumor radiosensitization. It is supplied as a research reagent with an associated product datasheet; lot-specific purity is provided on the certificate of analysis.

  • Potent and selective ATM kinase inhibitor suitable for DNA damage response studies.
  • Demonstrated activity in cellular and preclinical models, including radiosensitization.
  • Supplied with a product datasheet; certificate of analysis provides lot-specific purity.
  • Molecular weight 448.45 and molecular formula C23H21FN6O3.
  • CAS number 2020089-41-0; alternate registry entries have been reported in public databases.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.