Description
- Laryngeal Mirror designed for examining the larynx, pharynx, and oral structures during autopsy
- High-polish reflective mirror surface ensures clear, distortion-free visualization
- Ideal for documenting burn injury, foreign bodies, airway trauma, edema, and soot deposition
- Commonly used for fire deaths, strangulation cases, inhalation injury assessment, and airway pathology
- Stainless-steel handle provides stability and a firm grip while maneuvering inside the throat
- Available in multiple mirror diameters to accommodate various anatomical requirements
- Fully autoclavable and reusable; designed for repeated use in morgue and pathology settings
- Essential tool for forensic examiners, medical examiners, and pathology teaching labs
- Visual inspection of the larynx and vocal cords
- Evaluation of airway obstruction, aspiration, or edema
- Examination of thermal or chemical inhalation injuries
- Documentation of internal findings in medicolegal investigations
Specifications
Specifications
| Product Type | Mirror |
| Sterility | Non-sterile |
| Disposable | Reusable |