Description
- Set includes 12 total pieces:, 2 Fiber Optic Laryngoscope Handles, 5 Disposable Mac Blades (Sizes 15 depending on kit configuration), 5 Disposable Miller Blades (Sizes 04 depending on kit configuration)
- Bright fiber optic illumination for clear, shadow-free visualization
- Disposable design eliminates sterilization, preventing cross-contamination
- Mac curved blades for indirect epiglottic elevation; Miller straight blades for direct lifting
- Rigid, durable polymer blades maintain stability during intubation procedures
- Handles feature ergonomic, non-slip grip and standard fiber optic compatibility
- Ideal for anesthesia, EMS, critical care, transport bags, educational labs, and autopsy/postmortem airway access and examination
- Individually packaged blades ensure sterile, ready-to-use convenience
- Lightweight and balanced for precise control
Specifications
Specifications
| Product Type | Laryngoscope Set, Mac/Miller |
| Sterility | Non-sterile |
| Disposable | Single-use |