Description
- Complete 12-piece reusable fiber optic set includes: 2 Reusable Fiber Optic Handles, 5 Reusable Mac Blades (Sizes 15), 5 Reusable Miller Blades (Sizes 04)
- Bright fiber optic illumination provides clear, shadow-free airway visualization
- Premium stainless-steel blades designed for durability and long-term repeated use
- Fully autoclavable and corrosion-resistant for reliable sterilization
- Mac curved blades ideal for indirect epiglottic elevation
- Miller straight blades ideal for direct lifting—especially in pediatric or difficult airways
- Ergonomic handles with secure grip and standard fiber optic compatibility
- Suitable for anesthesia, emergency response, operating rooms, field kits, teaching labs, and autopsy/postmortem airway access and airway inspection
- Precision engineering ensures smooth connection, stable performance, and dependable illumination
Specifications
Specifications
| Product Type | Laryngoscope Set, Mac/Miller |
| Material | Fiber Optic |
| Sterility | Non-sterile |
| Disposable | Reusable |