Learn More
Surgical Design™ Laryngoscope Set of 5 - Fiber Optic Mac, Disposable
Shop All Surgical Design ProductsDescription
- Set includes:, 1 Fiber Optic Laryngoscope Handle, 4 Disposable Mac Blades (Sizes 1, 2, 3, 4)
- Fiber optic light channel delivers bright, uniform illumination for clear airway visualization
- Disposable blade design eliminates sterilization and cross-contamination concerns
- Mac curved blade profile for efficient epiglottis and glottic exposure
- Handle has a secure, anti-slip grip and is compatible with standard fiber optic Mac blades
- Durable blade construction allows reliable use during intubation or examination
- Ideal for anesthesia, emergency medicine, transport kits, military/field settings, and autopsy/postmortem airway access
- Individually packaged blades maintain sterility and readiness
- Lightweight configuration for easy handling and operation
Specifications
Specifications
| Product Type | Laryngoscope Set, Macintosh |
| Sterility | Non-sterile |
| Disposable | Single-use |