Learn More
Surgical Design™ Laryngoscope Set of 5, Fiber Optic Miller Reusable
Shop All Surgical Design ProductsDescription
- Set includes:, 1 Reusable Fiber Optic Laryngoscope Handle, 4 Reusable Miller Blades (Sizes 0, 1, 2, 3)
- Straight Miller blade design enables direct lifting of the epiglottis—ideal for pediatric and difficult airway management
- Fiber optic light channel offers bright, consistent illumination
- Premium stainless-steel blades engineered for long-term, repeated use
- Fully autoclavable for safe sterilization and reliable performance over time
- Handle features a secure, ergonomic grip and universal fiber optic compatibility
- Suitable for anesthesia, emergency setups, transport kits, operating rooms, and autopsy/postmortem airway access
- Corrosion-resistant construction withstands repeated cleaning and high-temperature cycles
- Provides excellent control for both standard and complex airway procedures
Specifications
Specifications
| Product Type | Laryngoscope Set, Miller |
| Material | Fiber Optic |
| Sterility | Non-sterile |
| Disposable | Reusable |