Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3B Scientific™ Larynx Chart
GREENER_CHOICESDPSCI_ED

Catalog No. S00265 Shop All 3B Scientific Products
Change view
Click to view available options
Dimensions (L x W):
50 x 67 cm (19.7 x 26.4 in.)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
S00265 50 x 67 cm (19.7 x 26.4 in.)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S00265 Supplier 3B Scientific™ Supplier No. 1001502
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The human larynx is displayed in full color and anatomical detail on this 3B Scientific chart. The anatomy of the larynx along with structure, function, and some diseases of the larynx are covered in this poster.

  • The human larynx is displayed in full color and anatomical detail on this chart.
  • The anatomy of the larynx along with structure, function, and some diseases of the larynx are covered in this poster.
  • This thickly laminated larynx anatomical chart is printed on premium glossy (200 g) UV resistant paper and comes with 2 sided lamination (75 micron).
  • The Larynx comes in a convenient poster size 50 x 67 cm (20 x 26 in.) and can be written on and wiped off with non permanent markers.
  • The 75 micron lamination ensures the anatomical chart does not curl up at the edges and the UV treatment ensures the chart does not get a faded yellow color over time.
  • The larynx anatomical chart is a great addition to any doctor's office or classroom.

Specifications

Dimensions (L x W) 50 x 67 cm (19.7 x 26.4 in.)
For Use With (Application) Doctor's Office or Classroom
Weight (English) 0.33 lb.
Product Type Larynx Chart
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.