Learn More
ERA WATERS CORP LAS MBAS
SDP
Catalog No. NC9675308
Encompass
MBAS Anionic Surfactants, One 10 mL Flame-Sealed Ampule containing 1,000 mg/L Linear Alkyl Benzene Sulfonate (LAS) in Water preserved with H2SO4 to be diluted and used as a calibration or calibration check standard for method 425.1
Catalog No. NC9675308 Supplier ERA WATERS CORP Supplier No. 975
MBAS Anionic Surfactants, One 10 mL Flame-Sealed Ampule containing 1,000 mg/L Linear Alkyl Benzene Sulfonate (LAS) in Water preserved with H2SO4 to be diluted and used as a calibration or calibration check standard for method 425.1