Learn More
Aviva Systems Biology Corp LAS1L Antibody - middle region (ARP34629_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34629_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LAS1L. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of Human LAS1L. Alternate names: Las1-like, dJ475B7.2ApplicationWB. Species reactivity: Cow, Horse, Human
Catalog No. 50-014-52969