Learn More
Medchemexpress LLC Lasalocid | 25999-31-9 | ≥98.0% | 25 MG
SDP
Catalog No. 5000413900
Encompass_Preferred
Lasalocid (Lasalocid-A) is an antibacterial and anticoccidial agent used as a feed additive. It also exhibits antitumor activity and is orally active.
  • Antibacterial agent
  • Anticoccidial agent
  • Can be used as feed additives
  • Exhibits antitumor activity
  • Orally active
Catalog No. 50-004-13900 Supplier Medchemexpress LLC Supplier No. 25MGHYB1071
Lasalocid (Lasalocid-A) is an antibacterial and anticoccidial agent used as a feed additive. It also exhibits antitumor activity and is orally active.
  • Antibacterial agent
  • Anticoccidial agent
  • Can be used as feed additives
  • Exhibits antitumor activity
  • Orally active