Learn More
TARGETMOL CHEMICALS INC Lasalocid A ammonium 1MG
SDP
Catalog No. 502878531
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Lasalocid A ammonium is an amine-selective ion carrier and feed additive with antimicrobial activity and antibiotic, growth-promoting properties, inhibits the protozoa of falciparum malaria, and can be used in the study of microbial infections. Purity 98.65%
Catalog No. 50-287-8531 Supplier TARGETMOL CHEMICALS INC Supplier No. T15717L1MG
Add to Cart
Add to Cart
Also available in 5 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Lasalocid A ammonium is an amine-selective ion carrier and feed additive with antimicrobial activity and antibiotic, growth-promoting properties, inhibits the protozoa of falciparum malaria, and can be used in the study of microbial infections. Purity 98.65%