Learn More
AdipoGen Lasalocid A sodium salt
SDP
Catalog No. 502560607
Encompass_Preferred
Chemical. CAS 25999-20-6. Formula C34H53NaO8. MW 612.8. Synthetic. Ionophore. Potent antibacterial agent and a coccidiostat. Able to form neutral complexes with monovalent and divalent cations and transport them through apolar phase including lipid bilayer membranes. It can transport also larger organic cations, like protonated dopamine.
Catalog No. 50-256-0607 Supplier AdipoGen Supplier No. CDXL0015M050
Chemical. CAS 25999-20-6. Formula C34H53NaO8. MW 612.8. Synthetic. Ionophore. Potent antibacterial agent and a coccidiostat. Able to form neutral complexes with monovalent and divalent cations and transport them through apolar phase including lipid bilayer membranes. It can transport also larger organic cations, like protonated dopamine.