Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lasalocid A Sodium Salt Analytical Standard, MilliporeSigma™ Supelco™

Catalog No. 111022766 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-2766 5 mg
11-102-2765 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-102-2766 Supplier MilliporeSigma Supelco Supplier No. 739965MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Lasalocid is an antibacterial agent, belonging to the carboxylic polyether group of antibiotics, produced by the fermentation of Streptomyces lasaliensis. It is commercially available as a sodium salt and is reported to be used as a coccidiostat in poultry.

TRUSTED_SUSTAINABILITY

Specifications

CAS 25999-20-6
Melting Point 175°C to 185°C; 180°C (decomposition) (literature)
Flash Point Not applicable
Molecular Formula C34H53NaO8
Linear Formula C34H53NaO8
Quantity 5 mg
MDL Number MFCD00078065
Synonym Lasalocid Sodium Salt
Optical Rotation [α]/D -28°C ± 3°C = 1 in. methanol
Formula Weight 612.77
Percent Purity ≥95% (HPLC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.