Learn More
Medchemexpress LLC Lasalocid (sodium) | 25999-20-6 | 99.0% | 1 ML
SDP
Catalog No. 5000414039
Encompass_Preferred
Lasalocid sodium (Lasalocid sodium-A) is an antibacterial and anticoccidial agent, often used as a feed additive. It also exhibits antitumor activity and is orally active. In cellular studies, it activates human PXR in HepG2 cells and inhibits liver stage *Plasmodium berghei* infection. It has demonstrated the ability to inhibit the growth of various bacteria, while selectively promoting beneficial bacteria in the rumen.
  • Acts as an antibacterial and anticoccidial agent
  • Exhibits antitumor activity
  • Orally active
  • Inhibits liver stage *Plasmodium berghei* infection
  • Induces apoptosis and autophagy in cancer cells
  • Arrests cell cycle at G0/G1 phase
  • Protects against *Pneumocystis carinii*-induced pneumonia
Catalog No. 50-004-14039 Supplier Medchemexpress LLC Supplier No. HYB1071A10MM/1ML
Lasalocid sodium (Lasalocid sodium-A) is an antibacterial and anticoccidial agent, often used as a feed additive. It also exhibits antitumor activity and is orally active. In cellular studies, it activates human PXR in HepG2 cells and inhibits liver stage *Plasmodium berghei* infection. It has demonstrated the ability to inhibit the growth of various bacteria, while selectively promoting beneficial bacteria in the rumen.
  • Acts as an antibacterial and anticoccidial agent
  • Exhibits antitumor activity
  • Orally active
  • Inhibits liver stage *Plasmodium berghei* infection
  • Induces apoptosis and autophagy in cancer cells
  • Arrests cell cycle at G0/G1 phase
  • Protects against *Pneumocystis carinii*-induced pneumonia