Learn More
Medchemexpress LLC Lasamide | 2736-23-4 | >99.9% | 270.09 | 100 G
SDP
Catalog No. 5000454599
Encompass_Preferred
Lasamide is a potent human Carbonic Anhydrase inhibitor identified with catalog number HY-W013316. It shows strong inhibitory activity against hCAXII and hCAIX, making it suitable for research applications targeting these enzymes.
  • Potent human Carbonic Anhydrase inhibitor
  • Ki value of 7.54 nM for hCAXII
  • Ki value of 2.67 nM for hCAIX
  • Available with purity exceeding 99%
  • Suitable for research use only
Catalog No. 50-004-54599 Supplier Medchemexpress LLC Supplier No. HYW013316100G
Lasamide is a potent human Carbonic Anhydrase inhibitor identified with catalog number HY-W013316. It shows strong inhibitory activity against hCAXII and hCAIX, making it suitable for research applications targeting these enzymes.
  • Potent human Carbonic Anhydrase inhibitor
  • Ki value of 7.54 nM for hCAXII
  • Ki value of 2.67 nM for hCAIX
  • Available with purity exceeding 99%
  • Suitable for research use only