Learn More
Cayman Chemical L-Ascorbic Acid 6-stearate, Purity: ≥95%, CAS: 10605-09-1, 100 mg
SDP
Catalog No. 502917261
Encompass_Preferred
An L-ascorbic acid and stearic acid derivative with antioxidant and anticancer activities; scavenges DPPH in a cell-free assay (IC50 = 3.31 µg/ml); LNPs encapsulating L-ascorbic acid 6-stearate induce apoptosis in HL-60 cells at 190 µM
Catalog No. 50-291-7261 Supplier Cayman Chemical Supplier No. 39142100 MG
An L-ascorbic acid and stearic acid derivative with antioxidant and anticancer activities; scavenges DPPH in a cell-free assay (IC50 = 3.31 µg/ml); LNPs encapsulating L-ascorbic acid 6-stearate induce apoptosis in HL-60 cells at 190 µM