Learn More
Med Vet International Jorvet Ceramic Laser Tip, 10mm x 0.8mm, J0941A, 5 Pack
Catalog No. 5000495557
Encompass_Preferred
For use with the AccuVet LX-20 CO2 Laser System. JorVet offers these commonly needed parts for the operation of the CO2 Laser. These parts are warranted to be identical in quality to any factory part. All laser tips are compatible with Luxar LXH-ST, LXH-TL, LXHA105, and LX-AH Handpieces. Non-sterile.
Catalog No. 50-004-95557 Supplier Med Vet International Supplier No. JOR941A
For use with the AccuVet LX-20 CO2 Laser System. JorVet offers these commonly needed parts for the operation of the CO2 Laser. These parts are warranted to be identical in quality to any factory part. All laser tips are compatible with Luxar LXH-ST, LXH-TL, LXHA105, and LX-AH Handpieces. Non-sterile.