Learn More
Glass Expansion Laser Ablation S13 Cup Adaptor
SDP
Catalog No. 502703172
Encompass_Preferred
Glass Ball Cup Adaptor FS13 with Aux. Port. Main port inlet has 8mm OD and 4mm ID, while Side Arm Aux. Port is 4mm OD and 2mm ID. Connect to 4mm and 8mm tubing using GAZ-04U and GAZ-08U, or GAZ-04 and GAZ-08.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-3172 Supplier Glass Expansion Supplier No. 218094140
Glass Ball Cup Adaptor FS13 with Aux. Port. Main port inlet has 8mm OD and 4mm ID, while Side Arm Aux. Port is 4mm OD and 2mm ID. Connect to 4mm and 8mm tubing using GAZ-04U and GAZ-08U, or GAZ-04 and GAZ-08.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001