Learn More
Chemglass Life Sciences Laser Cryo-Babies Labels, 0.5mL Tubes, 0.94" x 0.50", Orange, 2, 380/Pk
SDP
Catalog No. 501531661
Encompass_Preferred
Laser Cryo-Babies Labels, 0.5mL Tubes, 0.94" x 0.50", Orange, 2,380/Pk. Cryo-babies and Cryo-tags pre-cut chemically inert, polyolefin cryogenic storage labels resist most organic solvents and caustic agents. Labels will accept any marking instrument, and easily peel off backing sheet for application. They adhere to most plastics, glass, and metals without cracking, peeling, or degrading. Temperature-resistant from -196 to 100C, withstand boiling water baths, moderate oven temperatures (150C), autoclaving, and extreme freezer temperatures (liquid and vapor phase nitrogen). Label sheets are specially top-coated to readily accept laser printing. Heat-resistant, stay-flat sheets will not jam in laser printers. Ideally sized, they will fit a wide variety of cryovials, microtubes, storage boxes and other containers. Rainbow packs include four sheets of each color: blue, green, orange, red and yellow. Cryo-Tags and Cryo-Babies are a registered trademark of Diversified Biotech.
Catalog No. 50-153-1661 Supplier Chemglass Life Sciences Supplier No. CG4037222
Laser Cryo-Babies Labels, 0.5mL Tubes, 0.94" x 0.50", Orange, 2,380/Pk. Cryo-babies and Cryo-tags pre-cut chemically inert, polyolefin cryogenic storage labels resist most organic solvents and caustic agents. Labels will accept any marking instrument, and easily peel off backing sheet for application. They adhere to most plastics, glass, and metals without cracking, peeling, or degrading. Temperature-resistant from -196 to 100C, withstand boiling water baths, moderate oven temperatures (150C), autoclaving, and extreme freezer temperatures (liquid and vapor phase nitrogen). Label sheets are specially top-coated to readily accept laser printing. Heat-resistant, stay-flat sheets will not jam in laser printers. Ideally sized, they will fit a wide variety of cryovials, microtubes, storage boxes and other containers. Rainbow packs include four sheets of each color: blue, green, orange, red and yellow. Cryo-Tags and Cryo-Babies are a registered trademark of Diversified Biotech.