Learn More
Andwin Scientific LASER CRYO-TAGS 67mm x 25mm (2.64" x .98") Pack of 600
SDP
Supplier:  Andwin Scientific 91871258
Encompass_Preferred
Cryo-Tags can endure long term cryogenic storage conditions and conventional freezer storage. This product is chemically inert and is able to stick onto croygenic vials, microtubes, test tubes, glass vials, beakers and metal racks. The adhesive labels can also resist flash/snap freezing in liquid nitrogen, boiling water baths at 100C and dry heat up to 150C. This product is sold in a pack of 600 labels measuring 67mm x 25mm (2.64" x .98").
Catalog No. 50-821-011