Learn More
Electron Microscopy Sciences Laser Cryo-Tag 38 mm x 19 mm, Blue, General Purpose
SDP
Supplier:  Electron Microscopy Sciences 77564B
Encompass_Preferred
Labels for cryogenic storage are temperature-resistant from -196 degree C to 150 degree C. Cryo-Babies withstand freezer temperatures down to -196 degree C (liquid and vapor phase nitrogen), boiling water baths (100 degree C), autoclaving and moderate oven temperatures (150 degree C).
Tough - Chemically inert labels resist most organic solvents and caustic agents. They adhere to most plastics, glass and metals without cracking, peeling or degrading.
Label sheets are specially top-coated to readily accept laser printing. Heat-resistant, stay-flat sheets will not jam in laser printers.
Made with a flexible, polyolefin label material and acrylic adhesive. This unique combination expands and contracts together with temperature fluctuations resulting in a strong adhesive bond.
Color: Blue
Use: General Purpose
Catalog No. 50-321-92