Learn More
GA International Laser & Inkjet Matte Paper Labels 1.75" x 0.5" / 44.5mm x 12.7mm Rectangle Light Blue From -29°C to +120°C (-20°F to +248°F)
Catalog No. 5000571222
Encompass_Preferred
General use, high printout quality, matte paper labels for laser and inkjet printers. Ideal for office use on files, folders, boxes, and containers. Strong permanent adhesive will adhere to a variety of surfaces, including paper, plastic, glass, cardboard, wood, and metal. Free MS Word templates are available for easy formatting.
Catalog No. 50-005-71222 Supplier GA International Supplier No. LIP13LBL
General use, high printout quality, matte paper labels for laser and inkjet printers. Ideal for office use on files, folders, boxes, and containers. Strong permanent adhesive will adhere to a variety of surfaces, including paper, plastic, glass, cardboard, wood, and metal. Free MS Word templates are available for easy formatting.