Learn More
GA International Laser & Inkjet Matte Paper Labels 1.75" x 0.5" / 44.5mm x 12.7mm Rectangle Pink From -29°C to +120°C (-20°F to +248°F)
Catalog No. 5000572275
Encompass_Preferred
General use, high printout quality, matte paper labels for laser and inkjet printers. Ideal for office use on files, folders, boxes, and containers. Strong permanent adhesive will adhere to a variety of surfaces, including paper, plastic, glass, cardboard, wood, and metal. Free MS Word templates are available for easy formatting.
Catalog No. 50-005-72275 Supplier GA International Supplier No. LIP13PI
General use, high printout quality, matte paper labels for laser and inkjet printers. Ideal for office use on files, folders, boxes, and containers. Strong permanent adhesive will adhere to a variety of surfaces, including paper, plastic, glass, cardboard, wood, and metal. Free MS Word templates are available for easy formatting.