Learn More
GA International Laser & Inkjet Matte Paper Labels 4" x 3.33" / 101.6mm x 84.6mm Rectangle White From -29°C to +120°C (-20°F to +248°F)
Catalog No. 5000572274
Encompass_Preferred
General use, high printout quality, matte paper labels for laser and inkjet printers. Ideal for office use on files, folders, boxes, and containers. Strong permanent adhesive will adhere to a variety of surfaces, including paper, plastic, glass, cardboard, wood, and metal. Free MS Word templates are available for easy formatting.
Catalog No. 50-005-72274 Supplier GA International Supplier No. LIP15WH
General use, high printout quality, matte paper labels for laser and inkjet printers. Ideal for office use on files, folders, boxes, and containers. Strong permanent adhesive will adhere to a variety of surfaces, including paper, plastic, glass, cardboard, wood, and metal. Free MS Word templates are available for easy formatting.