Learn More
GA International Laser & Inkjet Matte Paper Labels 7.5" x 1.25" / 190.5mm x 31.8mm Rectangle White Ambient temperature
Supplier:  GA International LRP66WH
Encompass_Preferred
General use paper labels, with a strong permanent adhesive, for laser and inkjet printers. The matte labels will adhere to paper, glass, plastic, cardboard, metal, and even wood surfaces. Ideal for office use on files, shelves, drawers, boxes, containers, and as shipping labels.
Catalog No. 50-005-72745