Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC Tape Logic Laser Labels, Fluorescent Red, 1-3/4 in., 8000/Roll

Catalog No. 501913355
Encompass

Laser labels are designed for creating custom product or mailing labels. They are cost-effective, allow for on-the-fly customization, and can be applied to various smooth surfaces. This fluorescent laser label's bright color ensures important information is noticed.

  • Laser labels
  • Color: Fluorescent red
  • Material: Paper
  • Number of labels per roll: 8,000
  • For use with: Laser printers

Catalog No. 50-191-3355 Supplier MSC Supplier No. 39527965
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Laser labels are designed for creating custom product or mailing labels. They are cost-effective, allow for on-the-fly customization, and can be applied to various smooth surfaces. This fluorescent laser label's bright color ensures important information is noticed.

  • Laser labels
  • Color: Fluorescent red
  • Material: Paper
  • Number of labels per roll: 8,000
  • For use with: Laser printers

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.