Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC Tape Logic Laser Label, Pastel Blue, 2-5/8 in. Square, 3000/Roll

Catalog No. 501913313
Encompass

Laser labels are designed for creating custom product or mailing labels. They offer cost-effective, on-the-fly customization and can be used on various smooth surfaces such as paper, corrugated, wood, glass, plastic, and metal.

  • Create custom product or mailing labels
  • Cost-effective for on-the-fly customization
  • Adheres to a wide variety of smooth surfaces including paper, corrugated, wood, glass, plastic, and metal
  • For use with laser printers
  • Measures 2-5/8 in. in length
  • Measures 2-5/8 in. in width
  • Made of paper material
  • Pastel blue color
  • Contains 3,000 labels per roll

Catalog No. 50-191-3313 Supplier MSC Supplier No. 39527544
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Laser labels are designed for creating custom product or mailing labels. They offer cost-effective, on-the-fly customization and can be used on various smooth surfaces such as paper, corrugated, wood, glass, plastic, and metal.

  • Create custom product or mailing labels
  • Cost-effective for on-the-fly customization
  • Adheres to a wide variety of smooth surfaces including paper, corrugated, wood, glass, plastic, and metal
  • For use with laser printers
  • Measures 2-5/8 in. in length
  • Measures 2-5/8 in. in width
  • Made of paper material
  • Pastel blue color
  • Contains 3,000 labels per roll

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.