Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC Tape Logic Laser Label, White, 2-3/4 x 2-3/4 in., 900/Roll

Catalog No. 501913318
Encompass

Laser labels are used to create custom product or mailing labels. They allow for cost-effective, on-demand customization and can be used on various smooth surfaces including paper, corrugated, wood, glass, plastic, and metal.

  • White color
  • Contains 900 labels per roll
  • For use with laser printers
  • Measures 2-3/4 in. width
  • Measures 2-3/4 in. length
  • Paper material

Catalog No. 50-191-3318 Supplier MSC Supplier No. 39527593
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Laser labels are used to create custom product or mailing labels. They allow for cost-effective, on-demand customization and can be used on various smooth surfaces including paper, corrugated, wood, glass, plastic, and metal.

  • White color
  • Contains 900 labels per roll
  • For use with laser printers
  • Measures 2-3/4 in. width
  • Measures 2-3/4 in. length
  • Paper material

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.