Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC Tape Logic Laser Labels, White, 7.5 x 7.5 in, 500/Roll

Catalog No. 501917571
Encompass

Tape Logic laser labels are designed for use with laser printers. These white paper labels measure 7-1/2 in by 7-1/2 in and are supplied in a roll of 500.

  • Material: Paper
  • Color: White
  • Length: 7-1/2 in
  • Width: 7-1/2 in
  • Labels per roll: 500
  • For use with laser printers
  • Country of origin: United States

Catalog No. 50-191-7571 Supplier MSC Supplier No. 39571443
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tape Logic laser labels are designed for use with laser printers. These white paper labels measure 7-1/2 in by 7-1/2 in and are supplied in a roll of 500.

  • Material: Paper
  • Color: White
  • Length: 7-1/2 in
  • Width: 7-1/2 in
  • Labels per roll: 500
  • For use with laser printers
  • Country of origin: United States

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.